Assay Details
Target Gene Details
Entrez Gene ID: | 12630 |
Gene Name: | complement component factor i |
Gene Aliases: |
- |
Location: |
Chr.3:129835889-129875329 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cfi | NM_007686.2 | 12 | 1325 | NP_031712.2 |
AA530545.1 | 3 | 215 | ||
BC150751.1 | 12 | 1318 | ||
U47810.1 | 12 | 1328 | AAB00438.1 |
More Information
Set Membership: |
Intragenic Exonic |