Assay Details
Target Gene Details
Entrez Gene ID: | 11428 |
Gene Name: | aconitase 1 |
Gene Aliases: |
Aco-1, Irebp, Irp1 |
Location: |
Chr.4:40143265-40199009 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aco1 | NM_007386.2 | 21 | 3099 | NP_031412.2 |
AK027914.1 | 21 | 2886 | ||
AK085951.1 | 21 | 2849 | ||
AK129025.1 | 21 | 2881 | ||
AK163985.1 | 21 | 2898 | BAE37570.1 | |
BC005454.1 | 21 | 2859 | AAH05454.1 | |
X61147.1 | 21 | 2866 | CAA43455.1 |
More Information
Set Membership: |
Intragenic Exonic |