Assay Details
Target Gene Details
Entrez Gene ID: | 66860 |
Gene Name: | tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1 |
Gene Aliases: |
1200003E16Rik, Tanc, mKIAA1728 |
Location: |
Chr.2:59611848-59846213 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tanc1 | NM_001290659.1 | 7 | 877 | NP_001277588.1 |
NM_198294.3 | 8 | 1032 | NP_938036.2 | |
AK030022.1 | 5 | 636 | BAC26741.1 | |
BC079914.1 | 7 | 835 | ||
BC141367.1 | 7 | 867 | ||
BG295375.1 | 4 | 477 | ||
CN534596.1 |