Assay Details
Target Gene Details
Entrez Gene ID: | 230379 |
Gene Name: | alkaline ceramidase 2 |
Gene Aliases: |
2410116I05Rik, Asah3l, CRG-L1, maCER2 |
Location: |
Chr.4:86874342-86932490 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acer2 | NM_001290541.1 | 5 | 3483 | NP_001277470.1 |
NM_001290543.1 | NP_001277472.1 | |||
NM_139306.3 | 6 | 3621 | NP_647467.1 | |
AF282864.1 | 6 | 3580 | AAL40408.1 | |
AK035314.1 | 1 | 2517 | ||
AK085306.1 | BAC39416.1 | |||
BC059819.1 | 5 | 3483 | AAH59819.1 |