Assay Details
Target Gene Details
Entrez Gene ID: | 381489 |
Gene Name: | relaxin/insulin-like family peptide receptor 1 |
Gene Aliases: |
Gm1018, Lgr7 |
Location: |
Chr.3:79641600-79737951 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rxfp1 | NM_212452.1 | 16 | 1415 | NP_997617.1 |
AK137655.1 | 15 | 1396 | ||
AY509975.1 | 16 | 1415 | AAR97515.1 |
More Information
Set Membership: |
Intragenic Exonic |