Assay Details
Target Gene Details
Entrez Gene ID: | 80877 |
Gene Name: | LPS-responsive beige-like anchor |
Gene Aliases: |
C80285, D3Ertd775e, Lab300, Lba |
Location: |
Chr.3:86223568-86782705 on Build GRCm38 |
Assay Gene Location: | Within Intron 40 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lrba | NM_001077687.1 | NP_001071155.1 | ||
NM_001077688.1 | NP_001071156.1 | |||
NM_030695.2 | NP_109620.2 | |||
AF187731.1 | AAG14003.1 | |||
AF188506.1 | AAG15400.1 | |||
AF188507.1 | AAG15401.1 | |||
AK035483.1 | BAC29075.1 | |||
AK053299.1 |
Target Gene Details
Entrez Gene ID: | 23937 |
Gene Name: | mab-21-like 2 (C. elegans) |
Gene Aliases: |
- |
Location: |
Chr.3:86545581-86548283 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mab21l2 | NM_011839.3 | 1 | 1356 | NP_035969.2 |
AF126427.1 | 1 | 1702 | AAD39141.1 | |
AF149877.1 | 1 | 1098 | AAD40478.1 | |
AK053645.1 | 1 | 1357 | BAC35460.1 | |
AK144373.1 | 1 | 1521 | ||
BC046628.1 | 1 | 1322 | AAH46628.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |