Assay Details
Target Gene Details
Entrez Gene ID: | 72274 |
Gene Name: | RIKEN cDNA 1700027H10 gene |
Gene Aliases: |
- |
Location: |
Chr.3:45416584-45439309 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
1700027H10Rik | NR_040594.1 | |||
AK006417.1 |