Assay Details
Target Gene Details
Entrez Gene ID: | 229003 |
Gene Name: | helicase with zinc finger 2, transcriptional coactivator |
Gene Aliases: |
PDIP1, Pric285, mKIAA1769, mPDIP1 |
Location: |
Chr.2:181227615-181242058 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Helz2 | NM_183162.2 | 16 | 8415 | NP_898985.2 |
AB201714.1 | 15 | 8319 | ||
AK173259.1 | 3 | 5925 | BAD32537.1 | |
BC006779.1 | 8 | 1523 | AAH06779.1 | |
BC052532.1 | 1 | 138 | AAH52532.1 |
More Information
Set Membership: |
Intragenic Exonic |