Assay Details
Target Gene Details
Entrez Gene ID: | 17932 |
Gene Name: | myelin transcription factor 1 |
Gene Aliases: |
Nzf2, Nzf2a, Nzf2b, Nztf2 |
Location: |
Chr.2:181763332-181827781 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Myt1 | NM_001171615.1 | NP_001165086.1 | ||
NM_001171616.1 | 1 | 69 | NP_001165087.1 | |
NM_008665.4 | 1 | 69 | NP_032691.2 | |
AB082378.1 | 1 | 56 | BAC16512.1 | |
AK165786.1 | 1 | 85 | ||
BC063252.1 | AAH63252.1 | |||
CV559907.1 | 1 | 69 |
More Information
Set Membership: |
Intragenic Exonic |