Assay Details
Target Gene Details
Entrez Gene ID: | 16874 |
Gene Name: | LIM homeobox protein 6 |
Gene Aliases: |
Lhx6.1 |
Location: |
Chr.2:36081950-36105997 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lhx6 | NM_001083125.1 | 1 | 196 | NP_001076594.1 |
NM_008500.2 | 1 | 196 | NP_032526.2 | |
AJ000337.1 | 1 | 59 | CAA04011.1 | |
AK090339.2 | BAC41175.1 | |||
AK140987.1 | 1 | 197 |
More Information
Set Membership: |
Intragenic Exonic |