Assay Details
Target Gene Details
Entrez Gene ID: | 269252 |
Gene Name: | general transcription factor IIIC, polypeptide 4 |
Gene Aliases: |
5330400C03, AI426938, AU014771, AU017413, KAT12, TFIIIC90 |
Location: |
Chr.2:28822299-28840360 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gtf3c4 | NM_001166033.1 | 3 | 1502 | NP_001159505.1 |
NM_172977.3 | 2 | 1396 | NP_766565.2 | |
AK030351.1 | 2 | 1323 | BAC26916.1 | |
AK047698.1 | 1 | 506 | BAC33130.1 | |
AK141269.1 | 2 | 1377 | ||
AK146401.1 | 3 | 1458 | BAE27142.1 | |
AK146403.1 | 3 | 1458 | BAE27144.1 | |
AK150650.1 | 2 | 1378 | BAE29737.1 | |
AK151179.1 | 2 | 1378 | BAE30181.1 | |
AK153102.1 | 2 | 1378 | BAE31722.1 | |
BC061476.1 | AAH61476.1 | |||
BC095998.1 | 2 | 1335 | AAH95998.1 | |
BC117919.1 | 2 | 1308 | ||
BC117920.1 | 2 | 1306 |
More Information
Set Membership: |
Intragenic Exonic |