Assay Details
Target Gene Details
Entrez Gene ID: | 22283 |
Gene Name: | Usher syndrome 2A (autosomal recessive, mild) |
Gene Aliases: |
A930011D15Rik, A930037M10Rik, Gm676, Gm983, Mush2a, Usherin |
Location: |
Chr.1:188262017-188965497 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ush2a | NM_021408.3 | 12 | 2781 | NP_067383.3 |
AF151717.1 | 13 | 2810 | AAF70550.1 | |
DQ073638.1 | 12 | 2585 | AAZ23164.1 |