Assay Details
Target Gene Details
Entrez Gene ID: | 67839 |
Gene Name: | G-protein signalling modulator 1 (AGS3-like, C. elegans) |
Gene Aliases: |
1810037C22Rik, AW107933, Ags3 |
Location: |
Chr.2:26314606-26348237 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpsm1 | NM_001199146.1 | 13 | 2950 | NP_001186075.1 |
NM_001199147.1 | 14 | 3199 | NP_001186076.1 | |
NM_153410.5 | 14 | 3130 | NP_700459.2 | |
AK007593.1 | 1 | 224 | ||
AK048691.1 | 14 | 3124 | BAC33422.1 | |
AK075789.1 | 4 | 1703 | ||
AK082985.1 | 14 | 3074 | ||
AK084631.1 | 13 | 2892 | BAC39236.1 | |
AK211070.1 | 1 | 122 | ||
BC026486.1 | 7 | 1901 | AAH26486.1 | |
BC027004.1 | 11 | 2481 | AAH27004.1 | |
BC071197.1 | 14 | 3196 | AAH71197.1 |