Assay Details
Target Gene Details
Entrez Gene ID: | 381399 |
Gene Name: | BPI fold containing family B, member 4 |
Gene Aliases: |
Gm1006, Lplunc4 |
Location: |
Chr.2:153938217-153963852 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bpifb4 | NM_001034875.3 | 16 | 1886 | NP_001030047.2 |
AK143420.1 | 13 | 1250 | BAE25370.1 | |
BC147610.1 | 13 | 1249 | ||
BC147625.1 | 13 | 1249 |