Assay Details
Target Gene Details
Entrez Gene ID: | 19025 |
Gene Name: | cathepsin A |
Gene Aliases: |
AU019505, PPCA, Ppgb |
Location: |
Chr.2:164832872-164841032 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ctsa | NM_001038492.2 | 1 | 157 | NP_001033581.1 |
AK019861.1 | 1 | 115 | BAB31888.1 | |
BY319945.1 | 1 | 158 | ||
CX209432.1 | 1 | 94 | ||
J05261.1 | 1 | 45 | AAA39982.1 |
Target Gene Details
Entrez Gene ID: | 415115 |
Gene Name: | neuralized-like 2 (Drosophila) |
Gene Aliases: |
Neur2, Ozz, Ozz-E3 |
Location: |
Chr.2:164830730-164833596 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Neurl2 | NM_001082974.2 | 1 | 569 | NP_001076443.1 |
AK004524.1 | 1 | 430 | BAB23346.1 |
More Information
Set Membership: |
Intragenic Exonic |