Assay Details
Target Gene Details
Entrez Gene ID: | 27387 |
Gene Name: | SH2 domain containing 3C |
Gene Aliases: |
Chat, Nsp3, Shep1 |
Location: |
Chr.2:32721055-32755007 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sh2d3c | NM_001252547.1 | 4 | 361 | NP_001239476.1 |
NM_013781.3 | 5 | 847 | NP_038809.1 | |
NR_045535.1 | 6 | 1305 | ||
AB030442.1 | 4 | 324 | BAA90557.1 | |
AB043953.1 | 5 | 830 | BAA96361.1 | |
AF168364.1 | 5 | 788 | AAF13305.1 | |
AK042709.1 | 4 | 345 | BAC31340.1 | |
AK155165.1 | 5 | 783 | BAE33088.1 | |
AK172072.1 | 6 | 1226 | ||
BC113203.1 | 5 | 802 | AAI13204.1 | |
BY202561.1 |