Assay Details
Target Gene Details
Entrez Gene ID: | 27387 |
Gene Name: | SH2 domain containing 3C |
Gene Aliases: |
Chat, Nsp3, Shep1 |
Location: |
Chr.2:32721055-32755007 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sh2d3c | NM_013781.3 | 2 | 526 | NP_038809.1 |
NR_045535.1 | 2 | 526 | ||
AB043953.1 | 2 | 509 | BAA96361.1 | |
AF168364.1 | 2 | 467 | AAF13305.1 | |
AK155165.1 | 2 | 462 | BAE33088.1 | |
AK172072.1 | 2 | 447 | ||
BC113203.1 | 2 | 481 | AAI13204.1 |