Assay Details
Target Gene Details
Entrez Gene ID: | 14582 |
Gene Name: | growth factor independent 1B |
Gene Aliases: |
Gfi-1B |
Location: |
Chr.2:28609450-28621982 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gfi1b | NM_001160406.1 | 4 | 530 | NP_001153878.1 |
NM_008114.3 | 4 | 530 | NP_032140.1 | |
AF017275.1 | 4 | 489 | AAC40088.1 | |
AK157262.1 | 4 | 539 | BAE34016.1 | |
BC052654.1 | 4 | 505 | AAH52654.1 | |
BC132365.1 | 4 | 466 | ||
BC132367.1 | 4 | 466 | ||
BC145238.1 | 4 | 466 |
More Information
Set Membership: |
Intragenic Exonic |