Assay Details
Target Gene Details
Entrez Gene ID: | 394430 |
Gene Name: | UDP glycosyltransferase 1 family, polypeptide A10 |
Gene Aliases: |
A13, Ugt1a13 |
Location: |
Chr.1:88055411-88220002 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ugt1a10 | NM_201641.2 | NP_964003.2 | ||
AY227201.1 | AAP48600.1 | |||
BC089569.1 | AAH89569.1 |
Target Gene Details
Entrez Gene ID: | 394433 |
Gene Name: | UDP glucuronosyltransferase 1 family, polypeptide A5 |
Gene Aliases: |
- |
Location: |
Chr.1:88166012-88220002 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ugt1a5 | NM_201643.2 | 1 | 443 | NP_964005.2 |
AB094481.1 | 1 | 422 | BAD11305.1 | |
AY227196.1 | 1 | 443 | AAP48595.1 | |
BC139093.1 | 1 | 423 | ||
BC139094.1 | 1 | 423 |
Target Gene Details
Entrez Gene ID: | 394435 |
Gene Name: | UDP glucuronosyltransferase 1 family, polypeptide A6B |
Gene Aliases: |
A9', Ugt1a9 |
Location: |
Chr.1:88103257-88218998 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ugt1a6b | NM_201410.3 | NP_958812.3 | ||
AY227198.1 | AAP48597.1 | |||
BC026561.1 | AAH26561.1 |
Target Gene Details
Entrez Gene ID: | 394432 |
Gene Name: | UDP glucuronosyltransferase 1 family, polypeptide A7C |
Gene Aliases: |
A10', A13, Ugt1a10 |
Location: |
Chr.1:88095001-88220002 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ugt1a7c | NM_201642.4 | NP_964004.1 | ||
AK128918.1 | BAC87656.1 | |||
AK144599.1 | BAE25958.1 | |||
AK153157.1 | BAE31767.1 | |||
AK154973.1 | ||||
AK159496.1 | ||||
AK162317.1 | ||||
AK165465.1 | BAE38202.1 | |||
AY227199.1 | AAP48598.1 | |||
BC141205.1 | ||||
BC141206.1 |
Target Gene Details
Entrez Gene ID: | 394434 |
Gene Name: | UDP glucuronosyltransferase 1 family, polypeptide A9 |
Gene Aliases: |
UDPGT, UDPGT 1-9, UGT1-09, UGT1.9, UGTP4, Ugt1a12, mUGTBr/P |
Location: |
Chr.1:88070779-88220002 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ugt1a9 | NM_201644.2 | NP_964006.2 | ||
AY227200.1 | AAP48599.1 | |||
BC138699.1 | ||||
BC146021.1 | ||||
L27122.1 | AAA40524.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |