Assay Details
Target Gene Details
Entrez Gene ID: | 353156 |
Gene Name: | EGF-like domain 7 |
Gene Aliases: |
VE-statin, Zneu1 |
Location: |
Chr.2:26581028-26592682 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Egfl7 | NM_001164564.1 | 9 | 1032 | NP_001158036.1 |
NM_178444.4 | 10 | 1257 | NP_848538.3 | |
NM_198724.2 | 10 | 1256 | NP_942017.2 | |
NM_198725.2 | 10 | 1217 | NP_942018.2 | |
AF184973.1 | 10 | 1207 | AAF01322.1 | |
AK002601.1 | 10 | 1213 | BAB22222.1 | |
AK139377.1 | 9 | 1027 | BAE23983.1 | |
AK156158.1 | 10 | 1259 | ||
AW491662.1 | 1 | 149 | ||
AY239289.1 | 10 | 1218 | AAP69825.1 | |
AY239290.1 | 10 | 1240 | AAP69826.1 | |
AY309459.1 | 10 | 1230 | AAP74732.1 | |
BC024610.1 | 10 | 1240 | AAH24610.2 | |
BG292585.1 | 3 | 292 | ||
CF949270.1 | 2 | 293 |
More Information
Set Membership: |
Intragenic Exonic |