Assay Details
Target Gene Details
Entrez Gene ID: | 20452 |
Gene Name: | ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4 |
Gene Aliases: |
PST, PST-1, SIAT8-D, ST8SiaIV, Siat8d |
Location: |
Chr.1:95587682-95667649 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
St8sia4 | NM_001159745.1 | 4 | 935 | NP_001153217.1 |
NM_009183.2 | 4 | 935 | NP_033209.1 | |
AJ223956.1 | 4 | 926 | CAA11685.1 | |
AK003690.1 | 4 | 877 | ||
AK041723.1 | 4 | 868 | BAC31044.1 | |
BC060112.1 | 4 | 912 | AAH60112.1 |
More Information
Set Membership: |
Intragenic Exonic |