Assay Details
Target Gene Details
Entrez Gene ID: | 228829 |
Gene Name: | PHD finger protein 20 |
Gene Aliases: |
6820402O20Rik |
Location: |
Chr.2:156196647-156309953 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Phf20 | NM_172674.2 | 10 | 1559 | NP_766262.2 |
AK033017.1 | 9 | 1358 | BAC28129.1 | |
AK038573.1 | 10 | 1466 | BAC30050.2 | |
AK045309.1 | 10 | 1478 | BAC32304.1 | |
BC060121.1 | 10 | 1439 | AAH60121.1 |