Assay Details
Target Gene Details
Entrez Gene ID: | 271786 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 13 |
Gene Aliases: |
A230002A12, A230020F20, BB182356 |
Location: |
Chr.2:54436387-55118309 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Galnt13 | NM_173030.2 | 13 | 2867 | NP_766618.2 |
AK038492.1 | 1 | 683 | ||
BC131651.1 | 13 | 2546 | ||
BC131652.1 | 12 | 2577 |
More Information
Set Membership: |
Intragenic Exonic |