Assay Details
Target Gene Details
Entrez Gene ID: | 67856 |
Gene Name: | enoyl Coenzyme A hydratase domain containing 3 |
Gene Aliases: |
2310005D12Rik, AI662097 |
Location: |
Chr.2:6188465-6212994 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Echdc3 | NM_024208.4 | 5 | 1616 | NP_077170.2 |
AK009166.1 | 5 | 1617 | BAB26117.1 |