Assay Details
Target Gene Details
Entrez Gene ID: | 110074 |
Gene Name: | deoxyuridine triphosphatase |
Gene Aliases: |
5031412I06Rik, 5133400F09Rik, D2Bwg0749e, Dutp, dUTPase |
Location: |
Chr.2:125247248-125259049 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dut | NM_001159646.1 | 7 | 1420 | NP_001153118.1 |
NM_023595.6 | 6 | 1515 | NP_076084.2 | |
AK011407.1 | 6 | 1306 | BAB27599.1 | |
AK077244.1 | 6 | 1548 | BAC36706.1 | |
AK135438.1 | 7 | 1429 | BAE22532.1 | |
AK153285.1 | 6 | 1296 | BAE31868.1 |
More Information
Set Membership: |
Intragenic Exonic |