Assay Details
Target Gene Details
Entrez Gene ID: | 228026 |
Gene Name: | pyruvate dehydrogenase kinase, isoenzyme 1 |
Gene Aliases: |
B830012B01, D530020C15Rik |
Location: |
Chr.2:71873218-71903858 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdk1 | NM_172665.5 | 11 | 3249 | NP_766253.2 |
AK046805.1 | 11 | 3250 | BAC32879.1 | |
AK088599.1 | 3 | 2146 | BAC40444.1 | |
AK153649.1 | 11 | 3202 | BAE32134.1 | |
AK165090.1 | 1 | 656 | ||
BC027196.1 | 11 | 3172 | AAH27196.1 |
More Information
Set Membership: |
Intragenic Exonic |