Assay Details
Target Gene Details
Entrez Gene ID: | 70827 |
Gene Name: | trafficking protein, kinesin binding 2 |
Gene Aliases: |
2900022D04Rik, 4733401O11Rik, AI480836, AW046271, Als2cr3, CALS-C, GRIF-1, GRIF1, OIP98, mKIAA0549 |
Location: |
Chr.1:58900449-58973482 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trak2 | NM_172406.3 | 16 | 4380 | NP_765994.2 |
AK053848.1 | 1 | 1572 | BAC35555.2 | |
AK122305.1 | 6 | 2769 | BAC65587.1 | |
BC050027.1 | 3 | 2233 | AAH50027.1 | |
BC060681.1 | 15 | 4097 | AAH60681.1 |
More Information
Set Membership: |
Intragenic Exonic |