Assay Details
Target Gene Details
Entrez Gene ID: | 319266 |
Gene Name: | RIKEN cDNA A130010J15 gene |
Gene Aliases: |
- |
Location: |
Chr.1:193173469-193177832 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
A130010J15Rik | NM_001160359.1 | 2 | 1621 | NP_001153831.1 |
NM_001160360.1 | 2 | 1469 | NP_001153832.1 | |
NM_181048.3 | 2 | 1631 | NP_851391.1 | |
AK037364.1 | 2 | 1628 | BAC29794.1 | |
AK047686.1 | 2 | 1565 | BAC33124.1 | |
AK077339.1 | 2 | 1470 | BAC36759.1 | |
AK084448.1 | BAC39186.1 | |||
BC055955.1 | 2 | 1566 | AAH55955.1 |
Target Gene Details
Entrez Gene ID: | 215243 |
Gene Name: | TRAF3 interacting protein 3 |
Gene Aliases: |
6030423D04Rik, AI115021, T3jam |
Location: |
Chr.1:193175504-193201546 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Traf3ip3 | NM_153137.4 | 16 | 1954 | NP_694777.3 |
AK031400.1 | 16 | 1894 | BAC27388.1 | |
AY383616.1 | 14 | 1538 | AAQ84304.1 | |
BC137605.1 | 14 | 1565 | ||
BC171933.1 | 14 | 1565 |
More Information
Set Membership: |
Intragenic Exonic |