Assay Details
Target Gene Details
Entrez Gene ID: | 74019 |
Gene Name: | TRAF3 interacting protein 1 |
Gene Aliases: |
3930402D05Rik, AU041749, MIP-T3 |
Location: |
Chr.1:91494590-91529307 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Traf3ip1 | NM_028718.2 | 5 | 934 | NP_082994.1 |
AK014457.1 | 2 | 415 | BAB29364.1 | |
AY613437.1 | 5 | 937 | AAT72918.1 | |
BC117868.2 | 5 | 908 |