Assay Details
Target Gene Details
Entrez Gene ID: | 19281 |
Gene Name: | protein tyrosine phosphatase, receptor type, T |
Gene Aliases: |
R-PTP-T, RPTP-rfo, RPTP-rho, RPTPmam4, RPTPrho, mKIAA0283, mRPTPrho |
Location: |
Chr.2:161521988-162661147 on Build GRCm38 |
Assay Gene Location: | Within Exon 32 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprt | NM_001291149.1 | 31 | 4893 | NP_001278078.1 |
NM_001291150.1 | 31 | 4863 | NP_001278079.1 | |
NM_001291151.1 | 31 | 4860 | NP_001278080.1 | |
NM_021464.5 | 30 | 4833 | NP_067439.1 | |
AF244125.1 | 31 | 4834 | AAF44712.1 | |
AK220254.1 | 24 | 3443 | BAD90179.1 |
More Information
Set Membership: |
Intragenic Exonic |