Assay Details
Target Gene Details
Entrez Gene ID: | 381379 |
Gene Name: | mediator complex subunit 19 |
Gene Aliases: |
2410018M14Rik, 3110040A13Rik, LCMR1 |
Location: |
Chr.2:84678402-84688215 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Med19 | NM_025885.3 | 2 | 388 | NP_080161.1 |
AK005399.1 | 2 | 356 | BAC25114.1 | |
AK010552.1 | 2 | 366 | ||
AK160579.1 | 2 | 354 | BAE35886.1 | |
AK168787.1 | 2 | 356 | BAE40621.1 | |
AK201120.1 | 1 | 143 | ||
AK209282.1 | 2 | 156 | ||
BB611514.1 | 2 | 388 | ||
BC044207.1 | 2 | 250 | AAH44207.1 | |
BC082559.1 | 2 | 353 | AAH82559.1 |