Assay Details
Target Gene Details
Entrez Gene ID: | 19246 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 1 |
Gene Aliases: |
PTP-1B, PTP-HA2, PTP1B |
Location: |
Chr.2:167932303-167979385 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptpn1 | NM_011201.3 | 9 | 2729 | NP_035331.3 |
AK085713.1 | 1 | 1005 |
More Information
Set Membership: |
Intragenic Exonic |