Assay Details
Target Gene Details
Entrez Gene ID: | 57438 |
Gene Name: | membrane-associated ring finger (C3HC4) 7 |
Gene Aliases: |
Axo, Axot, Gtrgeo17 |
Location: |
Chr.2:60209892-60249211 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
March7 | NM_020575.2 | 5 | 1592 | NP_065600.1 |
AF155739.1 | 5 | 1592 | AAD38411.1 | |
AK143177.1 | 5 | 1612 | ||
AK166666.1 | 5 | 1599 | BAE38928.1 | |
AK167246.1 | 5 | 1574 | BAE39367.1 | |
AK168555.1 | 5 | 1595 | BAE40429.1 | |
BC025029.1 | 5 | 1556 | AAH25029.1 |
More Information
Set Membership: |
Intragenic Exonic |