Assay Details
Target Gene Details
Entrez Gene ID: | 228836 |
Gene Name: | discs, large homolog-associated protein 4 (Drosophila) |
Gene Aliases: |
AI225853, BC024558, DAP-4, DAP4, SAPAP-4, Sapap4, WBP16 |
Location: |
Chr.2:156613633-156764363 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dlgap4 | NM_001277186.1 | 3 | 704 | NP_001264115.1 |
NM_001277187.1 | 3 | 704 | NP_001264116.1 | |
NM_146128.6 | 3 | 704 | NP_666240.4 | |
AK122408.1 | 3 | 704 | BAC65690.2 | |
AY243849.2 | 1 | 284 | AAO89220.2 | |
BC141110.1 | 1 | 341 | ||
BC145394.1 | 1 | 341 | ||
BC145395.1 | 1 | 341 |
More Information
Set Membership: |
Intragenic Exonic |