Assay Details
Target Gene Details
Entrez Gene ID: | 17153 |
Gene Name: | myelin and lymphocyte protein, T cell differentiation protein |
Gene Aliases: |
Mpv17, Vip17 |
Location: |
Chr.2:127633226-127656695 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mal | NM_001171187.1 | 3 | 1393 | NP_001164658.1 |
NM_010762.5 | 4 | 1561 | NP_034892.1 | |
AK004639.1 | 4 | 1148 | BAB23430.1 | |
AK158653.1 | 4 | 1562 | BAE34599.1 | |
AK169172.1 | 4 | 1085 | BAE40951.1 | |
BC006826.1 | 4 | 1058 | AAH06826.1 | |
Y07626.1 | 4 | 1037 | CAA68907.1 |
More Information
Set Membership: |
Intragenic Exonic |