Assay Details
Target Gene Details
Entrez Gene ID: | 19124 |
Gene Name: | protein C receptor, endothelial |
Gene Aliases: |
AI325044, Ccca, Ccd41, Epcr |
Location: |
Chr.2:155751125-155755478 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Procr | NM_011171.2 | 3 | 695 | NP_035301.2 |
AK136541.1 | 3 | 555 | ||
AK143719.1 | 3 | 797 | BAE25514.1 | |
AK167302.1 | 3 | 635 | BAE39407.1 | |
BC028755.1 | 3 | 568 | AAH28755.1 | |
CT010175.1 | 3 | 514 | CAJ18383.1 | |
L39017.1 | 3 | 569 | AAC42049.1 | |
X63748.2 | 3 | 601 | CAA45281.2 |
More Information
Set Membership: |
Intragenic Exonic |