Assay Details
Target Gene Details
Entrez Gene ID: | 18347 |
Gene Name: | olfactory receptor 48 |
Gene Aliases: |
C3, IC3, MOR232-5 |
Location: |
Chr.2:89844066-89844971 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Olfr48 | NM_010990.1 | 1 | 453 | NP_035120.1 |
AF102522.1 | 1 | 279 | AAD13314.1 |
More Information
Set Membership: |
Intragenic Exonic |