Assay Details
Target Gene Details
Entrez Gene ID: | 71586 |
Gene Name: | interferon induced with helicase C domain 1 |
Gene Aliases: |
9130009C22Rik, Helicard, Hlcd, MDA5, RLR-2 |
Location: |
Chr.2:62595612-62646255 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ifih1 | NM_001164477.1 | 15 | 3293 | NP_001157949.1 |
NM_027835.3 | 16 | 3440 | NP_082111.2 | |
AF374384.1 | 16 | 3349 | AAM21359.1 | |
AK018602.1 | 5 | 774 | BAB31303.2 | |
AK153018.1 | 16 | 3347 | BAE31652.1 | |
AK153344.1 | 8 | 1374 | BAE31920.1 | |
AY075132.1 | AAL77205.1 | |||
BC004031.1 | 9 | 1582 | AAH04031.1 | |
BC019605.1 | 3 | 471 | AAH19605.1 | |
BC025508.1 | 11 | 1884 | AAH25508.1 | |
BC080200.1 | 15 | 3198 | AAH80200.1 | |
EF113547.1 | 16 | 3129 | ||
EF113548.1 | 16 | 3129 | ||
EU084750.1 | 16 | 3129 |