Assay Details
Target Gene Details
Entrez Gene ID: | 228094 |
Gene Name: | ceramide kinase-like |
Gene Aliases: |
Gm1958, Rp26 |
Location: |
Chr.2:79331105-79430520 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cerkl | NM_001048176.1 | 1 | 52 | NP_001041641.1 |
BY742285.1 | 1 | 62 | ||
HQ426680.1 | 1 | 52 | ||
HQ426681.1 | 1 | 52 | ||
HQ426682.1 | 1 | 52 | ||
HQ426683.1 | 1 | 52 | ||
HQ426684.1 | 1 | 52 | ||
HQ426685.1 | 1 | 52 | ||
HQ426686.1 | 1 | 52 | ||
HQ426687.1 | 1 | 52 | ||
HQ426688.1 | 1 | 52 | ||
HQ426689.1 | 1 | 52 | ||
HQ426690.1 | 1 | 52 |
More Information
Set Membership: |
Intragenic Exonic |