Assay Details
Target Gene Details
Entrez Gene ID: | 319513 |
Gene Name: | PC-esterase domain containing 1A |
Gene Aliases: |
A930025D01Rik, AI480484, Fam113a |
Location: |
Chr.2:130417684-130424641 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pced1a | NM_001114541.1 | 8 | 1824 | NP_001108013.1 |
NM_178762.4 | 7 | 2411 | NP_848877.2 | |
AK032354.1 | BAC27830.1 | |||
AK032762.1 | 7 | 2386 | BAC28010.1 | |
AK044590.1 | 4 | 3108 | BAC31990.1 | |
AK045248.1 | 6 | 2481 | BAC32279.1 | |
AK081584.1 | 5 | 1872 | BAC38264.1 | |
AK085613.1 | 6 | 2471 | BAC39486.1 | |
AK161998.1 |