Assay Details
Target Gene Details
Entrez Gene ID: | 228592 |
Gene Name: | RIKEN cDNA F830045P16 gene |
Gene Aliases: |
Sirpb3 |
Location: |
Chr.2:129457797-129536602 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
F830045P16Rik | NM_177653.3 | 4 | 966 | NP_808321.1 |
AK089892.1 | 4 | 967 | BAC40988.1 |
More Information
Set Membership: |
Intragenic Exonic |