Assay Details
Target Gene Details
Entrez Gene ID: | 73122 |
Gene Name: | transforming growth factor, beta receptor associated protein 1 |
Gene Aliases: |
3110018K12Rik, AU024090, AU042859, AW742551, Trap1 |
Location: |
Chr.1:43047265-43098622 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tgfbrap1 | NM_001013025.2 | 3 | 907 | NP_001013043.1 |
AK051333.1 | 2 | 2142 | BAC34606.1 | |
AK141742.1 | 2 | 463 | BAE24818.1 | |
AK159332.1 | 2 | 440 | BAE34997.1 | |
BC138866.1 | 3 | 821 | ||
BC138867.1 | 3 | 821 |