Assay Details
Target Gene Details
Entrez Gene ID: | 74748 |
Gene Name: | SLAM family member 8 |
Gene Aliases: |
5830408F06Rik, Blame, SBBI42 |
Location: |
Chr.1:172581377-172591191 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slamf8 | NM_029084.3 | 3 | 761 | NP_083360.2 |
AB196828.1 | 3 | 625 | ||
AB196829.1 | 3 | 625 | ||
AB196830.1 | 3 | 625 | ||
AB196831.1 | 3 | 625 | ||
AK017911.1 | 3 | 664 | BAB30999.1 | |
AK142633.1 | 3 | 762 | BAE25140.1 | |
AK156077.1 | 3 | 660 | BAE33575.1 | |
BC024587.1 | 3 | 681 | AAH24587.1 |
More Information
Set Membership: |
Intragenic Exonic |