Assay Details
Target Gene Details
Entrez Gene ID: | 226922 |
Gene Name: | potassium voltage-gated channel, subfamily Q, member 5 |
Gene Aliases: |
7730402H11, 9230107O05Rik, AA589396, D1Mgi1 |
Location: |
Chr.1:21398399-21962650 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kcnq5 | NM_001160139.1 | 1 | 128 | NP_001153611.1 |
NM_023872.3 | 1 | 128 | NP_076361.1 | |
AK039046.1 | ||||
AK147264.1 | 1 | 131 | ||
BE648243.1 |
More Information
Set Membership: |
Intragenic Exonic |