Assay Details
Target Gene Details
Entrez Gene ID: | 13603 |
Gene Name: | opsin 3 |
Gene Aliases: |
ERO, Ecpn |
Location: |
Chr.1:175662421-175692902 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Opn3 | NM_010098.3 | 3 | 914 | NP_034228.1 |
AF140241.1 | 3 | 872 | AAD32670.1 | |
AK043135.1 | 3 | 915 | BAC31471.1 | |
AK132822.1 | 3 | 851 | BAE21380.1 | |
BC104486.1 | 4 | 764 | AAI04487.1 | |
BC105646.1 | 4 | 711 | ||
BC105919.1 | 4 | 711 | AAI05920.1 | |
BC139805.1 | 4 | 712 |
More Information
Set Membership: |
Intragenic Exonic |