Assay Details
Target Gene Details
Entrez Gene ID: | 12633 |
Gene Name: | CASP8 and FADD-like apoptosis regulator |
Gene Aliases: |
2310024N18Rik, A430105C05Rik, CLARP, Cash, Casper, ENSMUSG00000072980, FLAME, FLAME-1, Flip, Gm9845, I-FLICE, MRIT, c-Flip |
Location: |
Chr.1:58711310-58759209 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cflar | NM_001289704.2 | NP_001276633.1 | ||
NM_001293804.1 | NP_001280733.1 | |||
NM_009805.4 | 1 | 64 | NP_033935.2 | |
NM_207653.5 | 1 | 64 | NP_997536.1 | |
AK009498.1 | 2 | 140 | ||
AK143812.1 | 1 | 57 | BAE25546.1 | |
AK152515.1 | ||||
AK161220.1 | 1 | 48 | BAE36245.1 | |
BB615039.1 | 1 | 60 | ||
BB856464.1 | 1 | 65 | ||
BC029223.1 | AAH29223.1 | |||
U97076.1 | AAC53281.1 |
More Information
Set Membership: |
Intragenic Exonic |