Assay Details
Target Gene Details
Entrez Gene ID: | 213452 |
Gene Name: | dual serine/threonine and tyrosine protein kinase |
Gene Aliases: |
A930019K20Rik, C430014H23Rik, C820013G01, Ripk5 |
Location: |
Chr.1:132417181-132466959 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dstyk | NM_172516.4 | 3 | 918 | NP_766104.2 |
AK020880.1 | 3 | 777 | BAB32238.1 | |
AK083696.1 | 3 | 816 | ||
AK132119.1 | 3 | 793 | ||
AK145449.1 | 3 | 763 | BAE26444.1 | |
AK172954.1 | 3 | 765 | BAD32232.1 | |
AK186755.1 | ||||
AK205064.1 | 1 | 91 | ||
AY208852.1 | 3 | 765 | AAP42420.1 | |
AY429676.1 | 3 | 804 | AAS55392.1 | |
BC130226.1 | 3 | 794 |
More Information
Set Membership: |
Intragenic Exonic |