Assay Details
Target Gene Details
Entrez Gene ID: | 435653 |
Gene Name: | Fc receptor-like B |
Gene Aliases: |
FREB-2, FREB2, FcRL2, Fcrlm2, Fcry, mFCRL2 |
Location: |
Chr.1:170906852-170913860 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fcrlb | NM_001029984.2 | 6 | 1212 | NP_001025155.2 |
AY513660.1 | 6 | 1212 | AAS82875.1 | |
DQ119292.1 | 6 | 1212 | AAZ23789.1 |
More Information
Set Membership: |
Intragenic Exonic |