Assay Details
Target Gene Details
Entrez Gene ID: | 98752 |
Gene Name: | Fc receptor-like A |
Gene Aliases: |
BB219290, FCRL, FCRL1, FREB, Fcrlm1, Fcrx, Freb1, mFREB, mFcrX |
Location: |
Chr.1:170917594-170927590 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fcrla | NM_001160215.1 | 3 | 355 | NP_001153687.1 |
NM_145141.2 | 3 | 352 | NP_660123.1 | |
NR_027666.1 | 3 | 421 | ||
AF329487.1 | 3 | 319 | AAL23897.1 | |
AF426462.1 | 3 | 312 | AAL58112.1 | |
AF531424.1 | 3 | 341 | AAM97592.1 | |
AK041572.1 | 3 | 332 | BAC30990.1 | |
AK172370.1 | 3 | 345 | BAE42968.1 | |
BB637253.1 | 3 | 348 | ||
BC064708.1 | 3 | 355 | AAH64708.1 | |
CJ080700.1 |
More Information
Set Membership: |
Intragenic Exonic |