Assay Details
Target Gene Details
Entrez Gene ID: | 78825 |
Gene Name: | desumoylating isopeptidase 2 |
Gene Aliases: |
5830417C01Rik, DeSI-2, Fam152a, Pppde1 |
Location: |
Chr.1:178187231-178252598 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Desi2 | NM_024282.3 | 5 | 2525 | NP_077244.1 |
AK020014.1 | 5 | 2410 | BAB31967.1 | |
AK029574.1 | BAC26520.1 | |||
AK146492.1 | 5 | 2258 | BAE27211.1 | |
BC046816.1 | 5 | 2232 | AAH46816.1 |
More Information
Set Membership: |
Intragenic Exonic |